Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDCD2
Entrez Gene ID
5134
Gene Name
programmed cell death 2
Gene Aliases
RP8, ZMYND7
Chromosome Location
Chr.6: 170575572 - 170584692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199462.1 NP_001186391.1 7 - 7 946 124
NM_002598.3 NP_002589.2 6 - 6 1083 124
GenBank mRNA AK055180.1 - 2 - 2 960 124
AY948416.1 - 7 - 7 873 124
BC008378.1 - 6 - 6 1045 124

View Full Product Details