The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199462.1 | NP_001186391.1 | 7 - 7 | 953 | 124 |
NM_002598.3 | NP_002589.2 | 6 - 6 | 1090 | 124 | |
GenBank mRNA | AK055180.1 | - | 2 - 2 | 967 | 124 |
AY948416.1 | - | 7 - 7 | 880 | 124 | |
BC008378.1 | - | 6 - 6 | 1052 | 124 |