The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_028062.1 | 1 - 2 | 502 | 89 | |
GenBank mRNA | BC074851.2 | - | 1 - 2 | 179 | 89 |
BC074852.2 | - | 1 - 2 | 179 | 89 | |
Y15801.1 | - | 1 - 2 | 501 | 89 |