The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145720.1 | NP_001139192.1 | 4 - 4 | 1519 | 99 |
GenBank mRNA | AL442095.1 | - | 2 - 2 | 306 | 99 |