Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPC1L1
Entrez Gene ID
29881
Gene Name
NPC1 like intracellular cholesterol transporter 1
Gene Aliases
NPC11L1
Chromosome Location
Chr.7: 44512440 - 44541650 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101648.1 NP_001095118.1 6 - 7 2221 126
NM_013389.2 NP_037521.2 6 - 7 2221 126
XM_011515327.2 XP_011513629.1 6 - 7 2556 126
XM_011515328.2 XP_011513630.1 3 - 4 1820 126
GenBank mRNA AF192522.1 - 6 - 7 2221 126
AY437865.1 - 6 - 7 2165 126
AY515256.1 - 6 - 7 2165 126
BC117178.1 - 6 - 7 2219 126
BC143756.1 - 6 - 7 2219 126
FJ481111.1 - 6 - 7 2165 126

View Full Product Details