The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139.2 | NP_001130.1 | 13 - 14 | 2018 | 109 |
GenBank mRNA | AF038461.1 | - | 13 - 14 | 2016 | 109 |
AF059250.1 | - | 13 - 14 | 1794 | 109 | |
BC041058.1 | - | 13 - 14 | 2046 | 109 |