Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALOX12B
Entrez Gene ID
242
Gene Name
arachidonate 12-lipoxygenase, 12R type
Gene Aliases
12R-LOX, ARCI2
Chromosome Location
Chr.17: 8072636 - 8087703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001139.2 NP_001130.1 4 - 5 787 98
GenBank mRNA AF038461.1 - 4 - 5 785 98
AF059250.1 - 4 - 5 563 98
BC041058.1 - 4 - 5 815 98

View Full Product Details