The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320747.1 | NP_001307676.1 | 17 - 18 | 2537 | 74 |
NM_001988.3 | NP_001979.2 | 17 - 18 | 2471 | 74 | |
GenBank mRNA | AK290333.1 | - | 17 - 18 | 2471 | 74 |
BC126103.1 | - | 17 - 18 | 2250 | 74 | |
BC126105.1 | - | 17 - 18 | 2250 | 74 | |
BC143814.1 | - | 17 - 18 | 2316 | 74 | |
U53786.1 | - | 17 - 18 | 2315 | 74 |