The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320747.1 | NP_001307676.1 | 20 - 21 | 2892 | 60 |
NM_001988.3 | NP_001979.2 | 20 - 21 | 2826 | 60 | |
GenBank mRNA | AK290333.1 | - | 20 - 21 | 2826 | 60 |
BC126103.1 | - | 20 - 21 | 2605 | 60 | |
BC126105.1 | - | 20 - 21 | 2605 | 60 | |
BC143814.1 | - | 20 - 21 | 2671 | 60 | |
U53786.1 | - | 20 - 21 | 2670 | 60 |