The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098509.1 | NP_001091979.1 | 20 - 21 | 2839 | 87 |
NM_014853.2 | NP_055668.2 | 21 - 22 | 2976 | 87 | |
XM_011524101.2 | XP_011522403.1 | 22 - 23 | 3052 | 87 | |
XM_011524102.2 | XP_011522404.1 | 22 - 23 | 3050 | 87 | |
XM_011524103.2 | XP_011522405.1 | 22 - 23 | 3051 | 87 | |
XM_011524104.2 | XP_011522406.1 | 22 - 23 | 3576 | 87 | |
XM_011524105.2 | XP_011522407.1 | 21 - 22 | 2916 | 87 | |
XM_011524106.1 | XP_011522408.1 | 21 - 22 | 2874 | 87 | |
XM_011524107.2 | XP_011522409.1 | 21 - 22 | 2870 | 87 | |
XM_011524108.1 | XP_011522410.1 | 20 - 21 | 2739 | 87 | |
XM_017025474.1 | XP_016880963.1 | 21 - 22 | 2914 | 87 | |
XM_017025475.1 | XP_016880964.1 | 21 - 22 | 2915 | 87 | |
XM_017025476.1 | XP_016880965.1 | 16 - 17 | 2211 | 87 | |
GenBank mRNA | AB007857.3 | - | 20 - 21 | 2839 | 87 |
AB275762.1 | - | 21 - 22 | 2811 | 87 | |
AB449878.1 | - | 20 - 21 | 2664 | 87 | |
AK294955.1 | - | 20 - 21 | 2731 | 87 | |
BC039204.1 | - | 21 - 22 | 2976 | 87 |