Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- AKR1C2
- Entrez Gene ID
-
1646
- Gene Name
- aldo-keto reductase family 1, member C2
- Gene Aliases
-
AKR1C-pseudo, BABP, DD, DD-2, DD/BABP, DD2, DDH2, HAKRD, HBAB, MCDR2, SRXY8, TDD
- Chromosome Location
-
Chr.10: 4987775 - 5018033 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_205845.2 |
NP_995317.1 |
1 - 2 |
134 |
92 |
GenBank mRNA |
BC063574.1 |
- |
1 - 2 |
101 |
92 |
View Full Product Details