The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256789.2 | NP_001243718.1 | 3 - 4 | 442 | 83 |
NM_001256790.2 | NP_001243719.1 | 3 - 4 | 247 | 83 | |
NM_005183.3 | NP_005174.2 | 3 - 4 | 442 | 83 | |
XM_011543983.2 | XP_011542285.1 | 3 - 4 | 280 | 83 | |
GenBank mRNA | AF067227.1 | - | 3 - 4 | 185 | 83 |
AF201304.1 | - | 3 - 4 | 442 | 83 | |
AJ224874.1 | - | 3 - 4 | 442 | 83 | |
JF701915.1 | - | 3 - 4 | 380 | 83 |