The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012469.3 | NP_036601.2 | 17 - 18 | 2507 | 71 |
XM_006723769.2 | XP_006723832.1 | 16 - 17 | 2297 | 71 | |
GenBank mRNA | AB019219.1 | - | 17 - 18 | 2421 | 71 |
AF026031.1 | - | 17 - 18 | 2438 | 71 | |
AF221842.1 | - | 17 - 18 | 2444 | 71 | |
AK001554.1 | - | 16 - 17 | 2330 | 71 | |
AK225993.1 | - | 16 - 17 | 2341 | 71 | |
AK314310.1 | - | 17 - 18 | 2431 | 71 | |
AL137320.1 | - | 8 - 9 | 1138 | 71 | |
BC001666.2 | - | 17 - 18 | 2385 | 71 |