Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRPF6
Entrez Gene ID
24148
Gene Name
pre-mRNA processing factor 6
Gene Aliases
ANT-1, ANT1, C20orf14, Prp6, RP60, SNRNP102, TOM, U5-102K, hPrp6
Chromosome Location
Chr.20: 63981069 - 64033100 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012469.3 NP_036601.2 18 - 19 2596 127
XM_006723769.2 XP_006723832.1 17 - 18 2386 127
GenBank mRNA AB019219.1 - 18 - 19 2510 127
AF026031.1 - 18 - 19 2527 127
AF221842.1 - 18 - 19 2533 127
AK001554.1 - 17 - 18 2419 127
AK225993.1 - 17 - 18 2430 127
AK314310.1 - 18 - 19 2520 127
AL137320.1 - 9 - 10 1227 127
BC001666.2 - 18 - 19 2474 127

View Full Product Details