Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDX27
Entrez Gene ID
55661
Gene Name
DEAD-box helicase 27
Gene Aliases
DRS1, Drs1p, HSPC259, PP3241, RHLP, dJ686N3.1
Chromosome Location
Chr.20: 49219295 - 49244077 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017895.7 NP_060365.7 6 - 7 756 100
XM_011528888.2 XP_011527190.1 6 - 7 727 100
GenBank mRNA AF193054.1 - 3 - 4 314 100
AK022979.1 - 6 - 7 626 100
AK025891.1 - 2 - 3 121 100
AK300135.1 - 6 - 7 635 100
AL832131.1 - 6 - 7 672 100
AY044431.1 - 6 - 7 695 100
BC009304.1 - 6 - 7 614 100
BC011927.2 - 6 - 7 610 100
BC016060.1 - 6 - 7 610 100
BC126287.1 - 6 - 7 745 100
BC130275.1 - 6 - 7 745 100
BC144125.1 - 6 - 7 745 100
EU718675.1 - 6 - 7 663 100

View Full Product Details