The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042574.2 | NP_001036039.1 | 5 - 6 | 667 | 59 |
NM_022769.4 | NP_073606.3 | 5 - 6 | 667 | 59 | |
XM_005254968.2 | XP_005255025.1 | 5 - 6 | 638 | 59 | |
XM_011521906.2 | XP_011520208.1 | 4 - 5 | 539 | 59 | |
GenBank mRNA | AK074170.1 | - | 2 - 3 | 138 | 59 |
AK090443.1 | - | 5 - 6 | 627 | 59 | |
AY360173.1 | - | 5 - 6 | 488 | 59 | |
BQ945777.1 | - | 5 - 6 | 616 | 59 |