The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014331.3 | NP_055146.1 | 8 - 9 | 1295 | 58 |
XM_011531802.2 | XP_011530104.1 | 8 - 9 | 1364 | 58 | |
GenBank mRNA | AB026891.1 | - | 8 - 9 | 1250 | 58 |
AB040875.1 | - | 8 - 9 | 1150 | 58 | |
AF200708.1 | - | 8 - 9 | 1260 | 58 | |
AF252872.1 | - | 8 - 9 | 1246 | 58 | |
AJ277882.1 | - | 8 - 9 | 1015 | 58 | |
AK290359.1 | - | 8 - 9 | 1295 | 58 | |
AK314855.1 | - | 8 - 9 | 1189 | 58 | |
BC012087.1 | - | 8 - 9 | 1175 | 58 |