The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002644.3 | NP_002635.2 | 1 - 2 | 133 | 60 |
GenBank mRNA | AF272149.2 | - | 1 - 2 | 129 | 60 |
S62403.1 | - | 1 - 2 | 105 | 60 | |
X73079.1 | - | 1 - 2 | 129 | 60 |