The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002644.3 | NP_002635.2 | 2 - 3 | 231 | 87 |
XM_011509629.1 | XP_011507931.1 | 2 - 3 | 142 | 87 | |
GenBank mRNA | AF272149.2 | - | 2 - 3 | 227 | 87 |
AK225471.1 | - | 2 - 3 | 231 | 87 | |
BC110494.2 | - | 2 - 3 | 128 | 87 | |
BC110495.2 | - | 2 - 3 | 128 | 87 | |
CR749533.1 | - | 3 - 4 | 304 | 87 | |
S62403.1 | - | 2 - 3 | 203 | 87 | |
X73079.1 | - | 2 - 3 | 227 | 87 |