The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052928.2 | NP_443160.2 | 4 - 5 | 534 | 82 |
XM_011523632.2 | XP_011521934.1 | 4 - 5 | 515 | 82 | |
XM_011523633.2 | XP_011521935.1 | 4 - 5 | 515 | 82 | |
XM_017024121.1 | XP_016879610.1 | 5 - 6 | 779 | 82 | |
GenBank mRNA | AK095369.1 | - | 4 - 5 | 534 | 82 |
BC035077.2 | - | 4 - 5 | 453 | 82 | |
DB219337.1 | - | 5 - 6 | 492 | 82 |