Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDHB13
Entrez Gene ID
56123
Gene Name
protocadherin beta 13
Gene Aliases
PCDH-BETA13
Chromosome Location
Chr.5: 141213919 - 141217421 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF217745.1 - 1 - 1 4809 68

View Full Product Details