The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012175.3 | NP_036307.2 | 2 - 3 | 228 | 132 |
NM_033406.2 | NP_208385.1 | 2 - 3 | 228 | 132 | |
XM_011519980.1 | XP_011518282.1 | 2 - 3 | 234 | 132 | |
XM_011519981.2 | XP_011518283.1 | 2 - 3 | 234 | 132 | |
XM_011519982.1 | XP_011518284.1 | 2 - 3 | 207 | 132 | |
XM_017017533.1 | XP_016873022.1 | 2 - 3 | 207 | 132 | |
GenBank mRNA | AB209039.1 | - | 1 - 2 | 3613 | 132 |
AF174595.1 | - | 2 - 3 | 188 | 132 | |
AF176702.1 | - | 2 - 3 | 185 | 132 | |
AK001943.1 | - | 2 - 3 | 219 | 132 | |
AK128454.1 | - | 2 - 3 | 206 | 132 | |
AK304866.1 | - | 2 - 3 | 223 | 132 | |
AL136625.1 | - | 2 - 3 | 211 | 132 | |
AL832826.1 | - | 1 - 2 | 900 | 132 | |
BC039291.1 | - | 2 - 3 | 228 | 132 | |
BC046110.1 | - | 2 - 3 | 214 | 132 | |
BG716664.1 | - | 2 - 3 | 231 | 132 |