The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017017059.1 | XP_016872548.1 | 1 - 2 | 114 | 94 |
XM_017017061.1 | XP_016872550.1 | 1 - 2 | 114 | 94 | |
XM_017017062.1 | XP_016872551.1 | 1 - 2 | 114 | 94 | |
GenBank mRNA | BM819966.1 | - | 1 - 2 | 114 | 94 |