The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039660.1 | NP_001034749.1 | 1 - 2 | 108 | 83 |
NM_173044.2 | NP_766632.2 | 1 - 2 | 108 | 83 | |
GenBank mRNA | AF215907.2 | - | 1 - 2 | 97 | 83 |
CB154498.1 | - | 1 - 2 | 111 | 83 |