Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TACC1
Entrez Gene ID
6867
Gene Name
transforming acidic coiled-coil containing protein 1
Gene Aliases
Ga55
Chromosome Location
Chr.8: 38728186 - 38853028 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006283.2 NP_006274.2 1 - 2 537 73
XM_005273625.4 XP_005273682.1 2 - 3 651 73
XM_011544631.1 XP_011542933.1 1 - 2 537 73
XM_011544632.2 XP_011542934.1 3 - 4 712 73
XM_011544633.1 XP_011542935.1 1 - 2 537 73
XM_011544634.1 XP_011542936.1 1 - 2 537 73
GenBank mRNA AF049910.1 - 1 - 2 478 73
AK304507.1 - 3 - 4 370 73
AK314620.1 - 1 - 2 537 73

View Full Product Details