Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TACC1
Entrez Gene ID
6867
Gene Name
transforming acidic coiled-coil containing protein 1
Gene Aliases
Ga55
Chromosome Location
Chr.8: 38728186 - 38853028 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146216.2 NP_001139688.1 2 - 3 601 94
NM_006283.2 NP_006274.2 2 - 3 655 94
XM_005273625.4 XP_005273682.1 3 - 4 769 94
XM_005273628.1 XP_005273685.1 2 - 3 276 94
XM_005273629.1 XP_005273686.1 3 - 4 289 94
XM_005273630.1 XP_005273687.1 2 - 3 297 94
XM_011544631.1 XP_011542933.1 2 - 3 655 94
XM_011544632.2 XP_011542934.1 4 - 5 830 94
XM_011544633.1 XP_011542935.1 2 - 3 655 94
XM_011544634.1 XP_011542936.1 2 - 3 655 94
XM_011544635.1 XP_011542937.1 2 - 3 359 94
XM_017013766.1 XP_016869255.1 2 - 3 276 94
XM_017013767.1 XP_016869256.1 3 - 4 289 94
XM_017013768.1 XP_016869257.1 2 - 3 276 94
XM_017013769.1 XP_016869258.1 3 - 4 289 94
XM_017013775.1 XP_016869264.1 2 - 3 297 94
XM_017013776.1 XP_016869265.1 3 - 4 289 94
GenBank mRNA AF049910.1 - 2 - 3 596 94
AK294931.1 - 2 - 3 256 94
AK295071.1 - 2 - 3 203 94
AK304507.1 - 4 - 5 488 94
AK314620.1 - 2 - 3 655 94
AY072876.2 - 2 - 3 207 94
BC041391.1 - 4 - 5 852 94
BX647696.1 - 2 - 3 603 94

View Full Product Details