Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYNC2H1
Entrez Gene ID
79659
Gene Name
dynein cytoplasmic 2 heavy chain 1
Gene Aliases
ATD3, DHC1b, DHC2, DNCH2, DYH1B, SRPS2B, SRTD3, hdhc11
Chromosome Location
Chr.11: 103109431 - 103479863 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080463.1 NP_001073932.1 4 - 5 760 135
NM_001377.2 NP_001368.2 4 - 5 760 135
XM_006718903.2 XP_006718966.1 4 - 5 760 135
XM_017018291.1 XP_016873780.1 4 - 5 760 135
XM_017018293.1 XP_016873782.1 4 - 5 760 135
GenBank mRNA AB231766.1 - 4 - 5 669 135
AB290167.1 - 4 - 5 814 135
AK125524.1 - 4 - 5 760 135

View Full Product Details