The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102396.1 | NP_001095866.1 | 2 - 3 | 364 | 81 |
NM_025073.2 | NP_079349.2 | 2 - 3 | 352 | 81 | |
NR_049741.1 | 2 - 3 | 414 | 81 | ||
GenBank mRNA | AK024821.1 | - | 2 - 3 | 342 | 81 |
AK126316.1 | - | 2 - 3 | 362 | 81 | |
AK225734.1 | - | 2 - 3 | 343 | 81 | |
BC005934.1 | - | 2 - 3 | 322 | 81 | |
CR749219.1 | - | 2 - 3 | 355 | 81 | |
CR936771.1 | - | 2 - 3 | 355 | 81 | |
DB459255.1 | - | 2 - 3 | 182 | 81 |