Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LPP
Entrez Gene ID
4026
Gene Name
LIM domain containing preferred translocation partner in lipoma
Gene Aliases
-
Chromosome Location
Chr.3: 188153050 - 188890671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167671.2 NP_001161143.1 3 - 4 378 105
NM_001167672.2 NP_001161144.1 2 - 3 347 105
NM_005578.4 NP_005569.1 3 - 4 435 105
XM_005247446.4 XP_005247503.1 3 - 4 412 105
XM_005247450.4 XP_005247507.1 2 - 3 381 105
XM_005247451.4 XP_005247508.1 3 - 4 412 105
XM_005247453.2 XP_005247510.1 2 - 3 384 105
XM_011512820.2 XP_011511122.1 5 - 6 679 105
XM_011512823.2 XP_011511125.1 2 - 3 2494 105
XM_011512827.2 XP_011511129.1 3 - 4 335 105
XM_011512828.2 XP_011511130.1 3 - 4 500 105
XM_011512831.2 XP_011511133.1 2 - 3 242 105
XM_011512833.2 XP_011511135.1 4 - 5 469 105
XM_011512834.2 XP_011511136.1 4 - 5 560 105
XM_017006377.1 XP_016861866.1 5 - 6 541 105
XM_017006378.1 XP_016861867.1 4 - 5 557 105
XM_017006379.1 XP_016861868.1 4 - 5 692 105
XM_017006380.1 XP_016861869.1 4 - 5 588 105
XM_017006381.1 XP_016861870.1 3 - 4 378 105
GenBank mRNA AK304035.1 - 2 - 3 289 105
AL833171.1 - 3 - 4 606 105
BC130584.1 - 2 - 3 237 105
BC144076.1 - 3 - 4 294 105
CD697098.1 - 2 - 3 367 105
CR457074.1 - 1 - 2 189 105
U49957.1 - 3 - 4 435 105

View Full Product Details