Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HUWE1
Entrez Gene ID
10075
Gene Name
HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
Gene Aliases
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1
Chromosome Location
Chr.X: 53532096 - 53686729 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031407.6 NP_113584.3 42 - 43 5928 68
XM_005261965.3 XP_005262022.1 41 - 42 5995 68
XM_011530747.1 XP_011529049.1 43 - 44 6171 68
XM_011530751.2 XP_011529053.1 42 - 43 6039 68
XM_017029191.1 XP_016884680.1 41 - 42 8923 68
XM_017029192.1 XP_016884681.1 41 - 42 8923 68
XM_017029193.1 XP_016884682.1 41 - 42 8902 68
XM_017029194.1 XP_016884683.1 41 - 42 8923 68
XM_017029195.1 XP_016884684.1 41 - 42 8923 68
XM_017029196.1 XP_016884685.1 41 - 42 8923 68
XM_017029197.1 XP_016884686.1 41 - 42 8923 68
XM_017029198.1 XP_016884687.1 42 - 43 6265 68
XM_017029199.1 XP_016884688.1 43 - 44 6192 68
XM_017029200.1 XP_016884689.1 42 - 43 6062 68
XM_017029201.1 XP_016884690.1 42 - 43 6079 68
XM_017029202.1 XP_016884691.1 42 - 43 6055 68
XM_017029203.1 XP_016884692.1 42 - 43 6061 68
XM_017029204.1 XP_016884693.1 40 - 41 8673 68
XM_017029205.1 XP_016884694.1 40 - 41 8653 68
XM_017029206.1 XP_016884695.1 41 - 42 8923 68
GenBank mRNA AB002310.3 - 11 - 12 2030 68
AB071605.1 - 15 - 16 2628 68
AY772009.1 - 40 - 41 5573 68
AY929612.1 - 39 - 40 5525 68
DQ097177.1 - 42 - 43 5927 68

View Full Product Details