Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HUWE1
Entrez Gene ID
10075
Gene Name
HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
Gene Aliases
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1
Chromosome Location
Chr.X: 53532096 - 53686729 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031407.6 NP_113584.3 47 - 48 6715 61
XM_005261965.3 XP_005262022.1 46 - 47 6782 61
XM_011530747.1 XP_011529049.1 48 - 49 6958 61
XM_011530751.2 XP_011529053.1 47 - 48 6826 61
XM_017029191.1 XP_016884680.1 46 - 47 9710 61
XM_017029192.1 XP_016884681.1 46 - 47 9707 61
XM_017029193.1 XP_016884682.1 46 - 47 9689 61
XM_017029194.1 XP_016884683.1 46 - 47 9710 61
XM_017029195.1 XP_016884684.1 46 - 47 9710 61
XM_017029196.1 XP_016884685.1 46 - 47 9707 61
XM_017029197.1 XP_016884686.1 46 - 47 9710 61
XM_017029198.1 XP_016884687.1 47 - 48 7052 61
XM_017029199.1 XP_016884688.1 48 - 49 6979 61
XM_017029200.1 XP_016884689.1 47 - 48 6849 61
XM_017029201.1 XP_016884690.1 47 - 48 6866 61
XM_017029202.1 XP_016884691.1 47 - 48 6842 61
XM_017029203.1 XP_016884692.1 47 - 48 6848 61
XM_017029204.1 XP_016884693.1 45 - 46 9460 61
XM_017029205.1 XP_016884694.1 45 - 46 9437 61
XM_017029206.1 XP_016884695.1 46 - 47 9710 61
GenBank mRNA AB002310.3 - 16 - 17 2814 61
AB071605.1 - 20 - 21 3415 61
AY772009.1 - 45 - 46 6360 61
AY929612.1 - 44 - 45 6312 61
DQ097177.1 - 47 - 48 6714 61

View Full Product Details