Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLXNC1
Entrez Gene ID
10154
Gene Name
plexin C1
Gene Aliases
CD232, PLXN-C1, VESPR
Chromosome Location
Chr.12: 94148696 - 94307675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005761.2 NP_005752.1 28 - 29 4638 93
NR_037687.1 9 - 10 1182 93
XM_006719186.3 XP_006719249.1 27 - 28 4626 93
GenBank mRNA AB208934.1 - 27 - 28 3319 93
AF030339.1 - 28 - 29 4638 93
AK295226.1 - 13 - 14 1767 93
AK304327.1 - 9 - 10 1182 93
AK307920.1 - 9 - 10 1085 93
AK308985.1 - 13 - 14 1407 93

View Full Product Details