Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GUF1
Entrez Gene ID
60558
Gene Name
GUF1 homolog, GTPase
Gene Aliases
EF-4
Chromosome Location
Chr.4: 44678395 - 44700685 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021927.2 NP_068746.2 13 - 14 1815 161
XM_005248122.3 XP_005248179.1 12 - 13 1669 161
XM_011513732.2 XP_011512034.1 13 - 14 1836 161
GenBank mRNA AK023282.1 - 13 - 14 1765 161
AK025248.1 - 5 - 6 595 161
AK225314.1 - 13 - 14 1770 161
BC036768.1 - 13 - 14 1776 161

View Full Product Details