The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015072.4 | NP_055887.3 | 12 - 13 | 1316 | 90 |
GenBank mRNA | AB023215.1 | - | 10 - 11 | 876 | 90 |
AK309748.1 | - | 12 - 13 | 1256 | 90 | |
AY237126.1 | - | 12 - 13 | 1246 | 90 | |
BC136472.1 | - | 12 - 13 | 1206 | 90 | |
BC136473.1 | - | 12 - 13 | 1206 | 90 |