The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015072.4 | NP_055887.3 | 18 - 19 | 1826 | 114 |
GenBank mRNA | AB023215.1 | - | 16 - 17 | 1386 | 114 |
AY237126.1 | - | 18 - 19 | 1756 | 114 | |
BC016625.1 | - | 5 - 6 | 407 | 114 | |
BC136472.1 | - | 18 - 19 | 1716 | 114 | |
BC136473.1 | - | 18 - 19 | 1716 | 114 | |
BX647530.1 | - | 11 - 12 | 857 | 114 |