The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015072.4 | NP_055887.3 | 31 - 32 | 4096 | 98 |
GenBank mRNA | AB023215.1 | - | 29 - 30 | 3656 | 98 |
AL133586.1 | - | 6 - 7 | 977 | 98 | |
AY237126.1 | - | 31 - 32 | 4026 | 98 | |
BC136472.1 | - | 31 - 32 | 3986 | 98 | |
BC136473.1 | - | 31 - 32 | 3986 | 98 |