Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELF2
Entrez Gene ID
1998
Gene Name
E74 like ETS transcription factor 2
Gene Aliases
EU32, NERF, NERF-1A, NERF-1B, NERF-1a,b, NERF-2
Chromosome Location
Chr.4: 139057717 - 139177218 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201999.2 NP_973728.1 1 - 2 67 155
XM_005262803.2 XP_005262860.1 2 - 3 340 155
XM_005262804.2 XP_005262861.1 2 - 3 340 155
XM_005262805.2 XP_005262862.1 2 - 3 340 155
XM_006714128.2 XP_006714191.1 2 - 3 340 155
XM_017007861.1 XP_016863350.1 1 - 2 88 155
GenBank mRNA AK311505.1 - 2 - 3 340 155
BQ221720.1 - 2 - 3 321 155
DR000047.1 - 1 - 2 67 155

View Full Product Details