The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000208.3 | NP_000199.2 | 19 - 20 | 3937 | 149 |
NM_001079817.2 | NP_001073285.1 | 18 - 19 | 3901 | 149 | |
XM_011527988.2 | XP_011526290.2 | 19 - 20 | 4037 | 149 | |
XM_011527989.2 | XP_011526291.2 | 18 - 19 | 4000 | 149 | |
GenBank mRNA | AB208861.1 | - | 11 - 12 | 1879 | 149 |
BC117172.1 | - | 18 - 19 | 3523 | 149 | |
M10051.1 | - | 19 - 20 | 3664 | 149 | |
X02160.1 | - | 18 - 19 | 3538 | 149 |