Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INSR
Entrez Gene ID
3643
Gene Name
insulin receptor
Gene Aliases
CD220, HHF5
Chromosome Location
Chr.19: 7112255 - 7294405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000208.3 NP_000199.2 3 - 4 1380 103
NM_001079817.2 NP_001073285.1 3 - 4 1380 103
XM_011527988.2 XP_011526290.2 3 - 4 1483 103
XM_011527989.2 XP_011526291.2 3 - 4 1482 103
GenBank mRNA BC047591.1 - 3 - 4 944 103
BC117172.1 - 3 - 4 1002 103
M10051.1 - 3 - 4 1107 103
X02160.1 - 3 - 4 1017 103

View Full Product Details