Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCOLN3
Entrez Gene ID
55283
Gene Name
mucolipin 3
Gene Aliases
TRP-ML3, TRPML3
Chromosome Location
Chr.1: 85018082 - 85048902 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253693.1 NP_001240622.1 8 - 9 1070 155
NM_018298.10 NP_060768.8 9 - 10 1238 155
XM_005271003.1 XP_005271060.1 10 - 11 1635 155
XM_006710750.1 XP_006710813.1 10 - 11 1454 155
XM_011541739.2 XP_011540041.2 9 - 10 1162 155
XM_011541740.2 XP_011540042.1 8 - 9 996 155
XM_011541742.1 XP_011540044.1 10 - 11 1372 155
GenBank mRNA AF475085.1 - 9 - 10 1144 155
AK001868.1 - 8 - 9 976 155
AK292206.1 - 9 - 10 1170 155
AK314354.1 - 9 - 10 1198 155

View Full Product Details