The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303007.1 | NP_001289936.1 | 2 - 3 | 1503 | 61 |
NM_001303008.1 | NP_001289937.1 | 3 - 4 | 603 | 61 | |
NM_013974.2 | NP_039268.1 | 3 - 4 | 678 | 61 | |
XM_011514448.2 | XP_011512750.1 | 3 - 4 | 754 | 61 | |
GenBank mRNA | AF070667.1 | - | 3 - 4 | 675 | 61 |
AF087894.1 | - | 3 - 4 | 675 | 61 | |
AK026191.1 | - | 2 - 3 | 1030 | 61 | |
AK098671.1 | - | 3 - 4 | 665 | 61 | |
BC001435.2 | - | 3 - 4 | 590 | 61 | |
EB387244.1 | - | 3 - 4 | 577 | 61 | |
FJ224290.1 | - | 3 - 4 | 675 | 61 |