Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMC3
Entrez Gene ID
219681
Gene Name
armadillo repeat containing 3
Gene Aliases
CT81, KU-CT-1
Chromosome Location
Chr.10: 22928024 - 23038527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282745.1 NP_001269674.1 13 - 14 1817 98
NM_001282746.1 NP_001269675.1 13 - 14 1817 98
NM_001282747.1 NP_001269676.1 10 - 11 1328 98
NM_173081.4 NP_775104.2 13 - 14 1817 98
XM_005252380.2 XP_005252437.1 12 - 13 1605 98
XM_005252381.2 XP_005252438.1 12 - 13 1638 98
XM_011519350.2 XP_011517652.1 13 - 14 1800 98
XM_011519351.2 XP_011517653.1 4 - 5 647 98
XM_017015831.1 XP_016871320.1 12 - 13 1605 98
XM_017015832.1 XP_016871321.1 13 - 14 1800 98
XM_017015833.1 XP_016871322.1 12 - 13 1605 98
GenBank mRNA AK098711.1 - 3 - 4 357 98
AK302104.1 - 10 - 11 1275 98
AK302985.1 - 13 - 14 1804 98
AY436643.1 - 13 - 14 1778 98
BC039312.1 - 13 - 14 1810 98

View Full Product Details