Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMC3
Entrez Gene ID
219681
Gene Name
armadillo repeat containing 3
Gene Aliases
CT81, KU-CT-1
Chromosome Location
Chr.10: 22928024 - 23038527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282746.1 NP_001269675.1 16 - 17 2131 68
XM_017015832.1 XP_016871321.1 16 - 17 2093 68
XM_017015833.1 XP_016871322.1 15 - 16 1919 68
GenBank mRNA AY436643.1 - 16 - 17 2092 68
BX115364.1 - 1 - 2 68

View Full Product Details