Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMC3
Entrez Gene ID
219681
Gene Name
armadillo repeat containing 3
Gene Aliases
CT81, KU-CT-1
Chromosome Location
Chr.10: 22928024 - 23038527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282745.1 NP_001269674.1 1 - 2 82 125
NM_001282746.1 NP_001269675.1 1 - 2 82 125
NM_173081.4 NP_775104.2 1 - 2 82 125
XM_005252380.2 XP_005252437.1 1 - 2 65 125
XM_011519350.2 XP_011517652.1 1 - 2 65 125
XM_017015831.1 XP_016871320.1 1 - 2 65 125
XM_017015832.1 XP_016871321.1 1 - 2 65 125
XM_017015833.1 XP_016871322.1 1 - 2 65 125
GenBank mRNA AK057389.1 - 1 - 2 25 125
AY436643.1 - 1 - 2 43 125
BC039312.1 - 1 - 2 75 125
DB459427.1 - 1 - 2 82 125

View Full Product Details