Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MROH1
Entrez Gene ID
727957
Gene Name
maestro heat like repeat family member 1
Gene Aliases
HEATR7A
Chromosome Location
Chr.8: 144148016 - 144261940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032450.2 NP_115826.2 23 - 24 2560 76
XM_011517270.2 XP_011515572.1 23 - 24 2698 76
XM_011517271.2 XP_011515573.1 23 - 24 2697 76
XM_011517273.2 XP_011515575.1 23 - 24 2696 76
XM_011517275.2 XP_011515577.1 23 - 24 2696 76
XM_011517276.2 XP_011515578.1 23 - 24 2724 76
XM_011517277.1 XP_011515579.1 23 - 24 2560 76
XM_011517278.1 XP_011515580.1 22 - 23 2440 76
XM_011517279.2 XP_011515581.1 22 - 23 2604 76
XM_011517281.1 XP_011515583.1 22 - 23 2613 76
XM_011517282.1 XP_011515584.1 21 - 22 2538 76
XM_011517283.2 XP_011515585.1 23 - 24 2519 76
XM_017013806.1 XP_016869295.1 20 - 21 2175 76
GenBank mRNA AL834309.1 - 22 - 23 2444 76

View Full Product Details