Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MROH1
Entrez Gene ID
727957
Gene Name
maestro heat like repeat family member 1
Gene Aliases
HEATR7A
Chromosome Location
Chr.8: 144148016 - 144261940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288814.1 NP_001275743.1 26 - 27 2860 70
NM_032450.2 NP_115826.2 28 - 29 3000 70
XM_011517270.2 XP_011515572.1 28 - 29 3138 70
XM_011517271.2 XP_011515573.1 28 - 29 3137 70
XM_011517272.2 XP_011515574.1 27 - 28 3111 70
XM_011517273.2 XP_011515575.1 28 - 29 3136 70
XM_011517274.2 XP_011515576.1 27 - 28 3109 70
XM_011517275.2 XP_011515577.1 28 - 29 3136 70
XM_011517276.2 XP_011515578.1 28 - 29 3164 70
XM_011517277.1 XP_011515579.1 28 - 29 3000 70
XM_011517278.1 XP_011515580.1 27 - 28 2880 70
XM_011517279.2 XP_011515581.1 27 - 28 3044 70
XM_011517281.1 XP_011515583.1 27 - 28 3053 70
XM_011517282.1 XP_011515584.1 26 - 27 2978 70
XM_011517283.2 XP_011515585.1 28 - 29 2959 70
XM_011517285.1 XP_011515587.1 26 - 27 2853 70
XM_017013802.1 XP_016869291.1 27 - 28 3109 70
XM_017013803.1 XP_016869292.1 27 - 28 3108 70
XM_017013804.1 XP_016869293.1 27 - 28 2973 70
XM_017013805.1 XP_016869294.1 26 - 27 2853 70
XM_017013806.1 XP_016869295.1 25 - 26 2615 70
GenBank mRNA AB058736.1 - 26 - 27 2865 70
AK295740.1 - 26 - 27 2853 70
AL834145.1 - 12 - 13 1371 70
AL834309.1 - 27 - 28 2884 70

View Full Product Details