Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MROH1
Entrez Gene ID
727957
Gene Name
maestro heat like repeat family member 1
Gene Aliases
HEATR7A
Chromosome Location
Chr.8: 144148016 - 144261940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099280.2 NP_001092750.1 6 - 7 699 93
NM_001099281.2 NP_001092751.1 5 - 6 579 93
NM_001288814.1 NP_001275743.1 5 - 6 586 93
NM_032450.2 NP_115826.2 6 - 7 699 93
XM_011517270.2 XP_011515572.1 6 - 7 837 93
XM_011517271.2 XP_011515573.1 6 - 7 836 93
XM_011517272.2 XP_011515574.1 6 - 7 837 93
XM_011517273.2 XP_011515575.1 6 - 7 835 93
XM_011517274.2 XP_011515576.1 6 - 7 835 93
XM_011517275.2 XP_011515577.1 6 - 7 835 93
XM_011517276.2 XP_011515578.1 6 - 7 863 93
XM_011517277.1 XP_011515579.1 6 - 7 699 93
XM_011517278.1 XP_011515580.1 5 - 6 579 93
XM_011517279.2 XP_011515581.1 5 - 6 743 93
XM_011517281.1 XP_011515583.1 5 - 6 752 93
XM_011517282.1 XP_011515584.1 4 - 5 677 93
XM_011517283.2 XP_011515585.1 6 - 7 658 93
XM_011517285.1 XP_011515587.1 5 - 6 579 93
XM_011517286.2 XP_011515588.1 6 - 7 831 93
XM_017013802.1 XP_016869291.1 6 - 7 835 93
XM_017013803.1 XP_016869292.1 6 - 7 834 93
XM_017013804.1 XP_016869293.1 6 - 7 699 93
XM_017013805.1 XP_016869294.1 5 - 6 579 93
XM_017013806.1 XP_016869295.1 3 - 4 314 93
XM_017013807.1 XP_016869296.1 6 - 7 832 93
GenBank mRNA AB058736.1 - 5 - 6 591 93
AK093563.1 - 6 - 7 698 93
AK295740.1 - 5 - 6 579 93
AL834309.1 - 5 - 6 583 93
BC040742.1 - 3 - 4 326 93
BC057824.1 - 6 - 7 679 93
BC067855.1 - 6 - 7 679 93
BC092486.1 - 5 - 6 559 93

View Full Product Details