Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMC3
Entrez Gene ID
219681
Gene Name
armadillo repeat containing 3
Gene Aliases
CT81, KU-CT-1
Chromosome Location
Chr.10: 22928024 - 23038527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282746.1 NP_001269675.1 14 - 15 1914 177
NM_001282747.1 NP_001269676.1 11 - 12 1425 177
NM_173081.4 NP_775104.2 14 - 15 1914 177
XM_005252380.2 XP_005252437.1 13 - 14 1702 177
XM_005252381.2 XP_005252438.1 13 - 14 1735 177
XM_011519350.2 XP_011517652.1 14 - 15 1897 177
XM_011519351.2 XP_011517653.1 5 - 6 744 177
XM_017015833.1 XP_016871322.1 13 - 14 1702 177
GenBank mRNA AK098711.1 - 4 - 5 454 177
AK302104.1 - 11 - 12 1372 177
AY436643.1 - 14 - 15 1875 177
BC039312.1 - 14 - 15 1907 177

View Full Product Details