The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039091.2 | NP_001034180.1 | 5 - 6 | 858 | 77 |
NM_002765.4 | NP_002756.1 | 5 - 6 | 849 | 77 | |
GenBank mRNA | AK313246.1 | - | 5 - 6 | 821 | 77 |
BC030019.2 | - | 5 - 6 | 790 | 77 | |
BC040483.1 | - | 5 - 6 | 792 | 77 | |
BC110875.1 | - | 5 - 6 | 783 | 77 | |
BC119662.1 | - | 5 - 6 | 790 | 77 | |
BC119663.1 | - | 5 - 6 | 781 | 77 | |
Y00971.1 | - | 5 - 6 | 766 | 77 |