The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004119.2 | NP_004110.2 | 10 - 11 | 1394 | 93 |
NR_130706.1 | 10 - 11 | 1394 | 93 | ||
XM_011535015.2 | XP_011533317.1 | 10 - 11 | 1393 | 93 | |
XM_011535017.2 | XP_011533319.1 | 10 - 11 | 1296 | 93 | |
XM_011535018.2 | XP_011533320.1 | 7 - 8 | 904 | 93 | |
XM_017020486.1 | XP_016875975.1 | 9 - 10 | 1173 | 93 | |
XM_017020487.1 | XP_016875976.1 | 11 - 12 | 1467 | 93 | |
XM_017020488.1 | XP_016875977.1 | 9 - 10 | 1160 | 93 | |
XM_017020489.1 | XP_016875978.1 | 9 - 10 | 1255 | 93 | |
GenBank mRNA | BC036028.1 | - | 10 - 11 | 1394 | 93 |
BC126350.1 | - | 10 - 11 | 1378 | 93 | |
BC144039.1 | - | 10 - 11 | 1379 | 93 | |
BC144040.1 | - | 10 - 11 | 1379 | 93 | |
U02687.1 | - | 10 - 11 | 1369 | 93 | |
Z26652.1 | - | 10 - 11 | 1312 | 93 |